Anti-TMEM132B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91364
102217-266EA 564.74 USD
102217-266
Anti-TMEM132B Rabbit Polyclonal Antibody
Antibodies
The TMEM132B Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM132B. This antibody reacts with human. The TMEM132B Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TMEM132B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR