Anti-TMEM110 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69238
102198-728EA 564.74 USD
102198-728
Anti-TMEM110 Rabbit Polyclonal Antibody
Antibodies
The TMEM110 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM110. This antibody reacts with human. The TMEM110 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TMEM110
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR