Anti-TMCC1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59916
102193-512EA 564.74 USD
102193-512
Anti-TMCC1 Rabbit Polyclonal Antibody
Antibodies
The TMCC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMCC1. This antibody reacts with human. The TMCC1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TMCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR