|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | TM9SF1 |
Antigen Name | Transmembrane 9 Superfamily Member 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
Antigen Synonyms | MP70 protein family member, transmembrane protein 9 superfamily member 1, multispanning membrane protein (70kD), HMP70, MP70, transmembrane 9 superfamily member 1 |
Gene ID | 10548 |
Immunogen | Synthetic peptides corresponding to TM9SF1(transmembrane 9 superfamily member 1) The peptide sequence was selected from the N terminal of TM9SF1. Peptide sequence YMEESGFLPHSHKIGLWTHLDFHLEFHGDRIIFANVSVRDVKPHSLDGLR. |
Purification | Immunogen affinity purified |
Storage Buffer | PBS and 2% Sucrose |
Storage Temperature | Store at -20C. Avoid freeze-thaw cycles. |