Anti-TLR9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77254
102211-194EA 564.74 USD
102211-194
Anti-TLR9 Rabbit Polyclonal Antibody
Antibodies
The TLR9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR9. This antibody reacts with human, mouse. The TLR9 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TLR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR