|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | TLR7 |
Antigen Name | Toll-like Receptor 7 |
Reactivity | Human, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
Antigen Synonyms | toll-like receptor 7 |
Gene ID | 51284 |
Concentration | 1.0 mg/ml |
Immunogen | Rabbit anti-TLR7 (IN) polyclonal antibody was raised against a peptide corresponding to 14 amino acids near the middle of mouse TLR7 (GenBank accession no. NP_573474). |
Purification | IgG purified |
Storage Buffer | PBS |
Storage Temperature | Store at 4C. Do not freeze. |