Anti-TLR6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54336
102183-944EA 564.74 USD
102183-944
Anti-TLR6 Rabbit Polyclonal Antibody
Antibodies
The TLR6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR6. This antibody reacts with human. The TLR6 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR