Anti-TLR6 Rabbit Polyclonal Antibody

NB110-93544 NB110-93544SS
102126-308EA 112.31 USD
102126-308 102126-306
Anti-TLR6 Rabbit Polyclonal Antibody
The TLR6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR6. This antibody reacts with human, mouse, rat, primate. The TLR6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR