Anti-TLR3 Rabbit Polyclonal Antibody

NB110-93591
102126-310EA 352.31 USD
102126-310
Anti-TLR3 Rabbit Polyclonal Antibody
The TLR3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR3. This antibody reacts with human, porcine, feline, primate. The TLR3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TLR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Porcine, Feline, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR