Anti-TLR2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77229
102211-102EA 536.43 USD
102211-102
Anti-TLR2 Rabbit Polyclonal Antibody
Antibodies
The TLR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR2. This antibody reacts with human, mouse. The TLR2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: TLR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR