Anti-TLR2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56720
102110-264EA 491.08 USD
102110-264
Anti-TLR2 Rabbit Polyclonal Antibody
Antibodies
The TLR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR2. This antibody reacts with human, mouse. The TLR2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: TLR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR