Anti-TLR11 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77204
102211-004EA 564.74 USD
102211-004
Anti-TLR11 Rabbit Polyclonal Antibody
Antibodies
The TLR11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR11. This antibody reacts with mouse. The TLR11 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: TLR11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR