Anti-TLR1 Rabbit Polyclonal Antibody

NBP1-77244
102211-160EA 466.15 USD
102211-160
Anti-TLR1 Rabbit Polyclonal Antibody
The TLR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR1. This antibody reacts with human, mouse. The TLR1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: TLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR