Anti-TKTL1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31674
102164-972EA 635.51 USD
102164-972
Anti-TKTL1 Rabbit Polyclonal Antibody
Antibodies
The TKTL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TKTL1. This antibody reacts with human. The TKTL1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: TKTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR