Anti-PLIN3 Rabbit Polyclonal Antibody

NBP1-76718
102209-096EA 466.15 USD
102209-096
Anti-PLIN3 Rabbit Polyclonal Antibody
The Perilipin-3 / TIP47 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Perilipin-3 / TIP47. This antibody reacts with human, rat. The Perilipin-3 / TIP47 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLIN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR