Anti-PLIN3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB110-40764B
102122-074EA 614.15 USD
102122-074
Anti-PLIN3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
The Perilipin-3 / TIP47 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to Perilipin-3 / TIP47. This antibody reacts with human, mouse, porcine, primate. The Perilipin-3 / TIP47 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: PLIN3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Porcine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR