Anti-TIP120B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78048
102212-420EA 449 USD
102212-420
Anti-TIP120B Rabbit Polyclonal Antibody
Antibodies
The TIP120B Antibody from Novus Biologicals is a rabbit polyclonal antibody to TIP120B. This antibody reacts with human. The TIP120B Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TIP120B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR