Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Tie-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69753SS NBP1-69753
The Tie-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Tie-2. This antibody reacts with human, mouse. The Tie-2 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Tie-2
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.025 ml NBP1-69753SS 102199-346 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

100 μl NBP1-69753 102199-344 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol Tie-2
Antigen Name TEK receptor tyrosine kinase
Reactivity Mouse, Human Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms angiopoietin-1 receptor, endothelial, Tunica interna endothelial cell kinase, EC 2.7.10, CD202b antigen, p140 TEK, Tyrosine-protein kinase receptor TEK, hTIE2, TIE2CD202b, TIE-2, CD202B, venous malformations, VMCM, soluble TIE2 variant 1, VMCM1, EC, multiple cutaneous and mucosal, TEK tyrosine kinase, soluble TIE2 variant 2, Tyrosine-protein kinase receptor TIE-2 multiple cutaneous and mucosal, Tyrosine-protein kinase receptor TIE-2, CD202b antigen, EC 2.7.10, p140 TEK, soluble TIE2 variant 2, TIE-2, TEK tyrosine kinase, VMCM1, soluble TIE2 variant 1, endothelial, angiopoietin-1 receptor, Tunica interna endothelial cell kinase, EC, Tyrosine-protein kinase receptor TEK, TIE2CD202b, venous malformations, VMCM, CD202B, hTIE2
Gene ID 7010
Concentration 1.0 mg/ml
Immunogen A genomic peptide made to an internal region of the human TIE2 protein (within residues 250-400). [Swiss-Prot Q02763]
Purification Immunogen affinity purified
Storage Buffer PBS
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Tested Applications Simple Western,
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty