Rabbit Polyclonal anti-Thyroid Stimulating Hormone Antibody, Novus Biologicals (NBP1-36643)

NBP1366430.5ML NBP1366437.0ML NBP1366430.1ML NBP1366431.0ML
102168-586EA 466.15 USD
102168-586 102168-584 102168-582 102168-580
Rabbit Polyclonal anti-Thyroid Stimulating Hormone Antibody, Novus Biologicals (NBP1-36643)
Rabbit Polyclonal Thyroid Stimulating Hormone Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: Thyroid Stimulating Hormone
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR