Anti-Thyroid receptor-interacting protein 13 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-04045
102234-850EA 635.51 USD
102234-850
Anti-Thyroid receptor-interacting protein 13 Rabbit Polyclonal Antibody
Antibodies
The Thyroid receptor-interacting protein 13 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Thyroid receptor-interacting protein 13. This antibody reacts with human. The Thyroid receptor-interacting protein 13 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: Thyroid receptor-interacting protein 13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR