Anti-thymocyte selection-associated high mobility group box Rabbit Polyclonal Antibody

NBP1-80030
102215-708EA 466.15 USD
102215-708
Anti-thymocyte selection-associated high mobility group box Rabbit Polyclonal Antibody
The thymocyte selection-associated high mobility group box Antibody from Novus Biologicals is a rabbit polyclonal antibody to thymocyte selection-associated high mobility group box. This antibody reacts with human. The thymocyte selection-associated high mobility group box Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: thymocyte selection-associated high mobility group box
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR