Anti-Thrombopoietin/Tpo Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59290
102192-282EA 564.74 USD
102192-282
Anti-Thrombopoietin/Tpo Rabbit Polyclonal Antibody
Antibodies
The Thrombopoietin / Tpo Antibody from Novus Biologicals is a rabbit polyclonal antibody to Thrombopoietin / Tpo. This antibody reacts with human. The Thrombopoietin / Tpo Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Thrombopoietin/Tpo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR