Anti-THOP1 Rabbit Polyclonal Antibody

NBP1-31547
102164-798EA 493.85 USD
102164-798
Anti-THOP1 Rabbit Polyclonal Antibody
The Thimet Oligopeptidase / THOP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Thimet Oligopeptidase / THOP1. This antibody reacts with human. The Thimet Oligopeptidase / THOP1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: THOP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR