Anti-THG1L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74219
102205-180EA 564.74 USD
102205-180
Anti-THG1L Rabbit Polyclonal Antibody
Antibodies
The THG1L Antibody from Novus Biologicals is a rabbit polyclonal antibody to THG1L. This antibody reacts with mouse. The THG1L Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: THG1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR