Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGN46 Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NBP1-49643R
The TGN46 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to TGN46. This antibody reacts with human. The TGN46 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: TGN46
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-49643R 102179-342 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TGN46
Antigen Name Trans-Golgi Network Integral Membrane Protein 2
Reactivity Human
Conjugation DyLight® 550
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms TGN48, TTGN 2, TGN-46, trans golgi network 38, TGN 51, TGN38 homolog, Trans-Golgi network protein TGN51, Trans Golgi network protein (46 48 51kD isoforms), TGOLN2, Trans-Golgi network integral membrane protein 2, TGN38, Trans Golgi network protein TGN51, Trans golgi network protein 2, trans golgi network 46, TGN51, TGOLN 2, TGN 48, Trans Golgi network integral membrane protein 2 [Precursor], Trans-Golgi network protein 2, TGN 46, TGN46, TTGN2, TGN-38, Trans Golgi network integral membrane protein 2
Gene ID 10618
Immunogen A genomic peptide made to an internal region of the human TGN46 protein (within residues 200-350). [Swiss-Prot O43493]
Purification Immunogen affinity purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping