Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-TGN46 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
The TGN46 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to TGN46. This antibody reacts with human. The TGN46 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: TGN46
Clonality: Polyclonal
Conjugation: DyLight 488
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-49643G 102179-338 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol TGN46
Antigen Name Trans-Golgi Network Integral Membrane Protein 2
Reactivity Human
Conjugation DyLight® 488
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms Trans Golgi network protein TGN51, TGOLN 2, Trans Golgi network integral membrane protein 2 [Precursor], TGN51, trans golgi network 46, TGN 46, TGN-38, TTGN2, TGN 51, TGN46, Trans Golgi network integral membrane protein 2, TGOLN2, TGN38 homolog, Trans-Golgi network protein 2, TGN48, TGN 48, Trans-Golgi network integral membrane protein 2, Trans-Golgi network protein TGN51, TGN38, Trans Golgi network protein (46 48 51kD isoforms), TGN-46, trans golgi network 38, TTGN 2, Trans golgi network protein 2
Gene ID 10618
Immunogen A genomic peptide made to an internal region of the human TGN46 protein (within residues 200-350). [Swiss-Prot O43493]
Purification Immunogen affinity purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty