Rabbit Polyclonal anti-TGF beta 2 Antibody, Novus Biologicals (NBP1-36573)

NBP1365730.1ML NBP1365730.5ML NBP1365731.0ML NBP1365737.0ML
102168-538EA 466.15 USD
102168-538 102168-536 102168-542 102168-540
Rabbit Polyclonal anti-TGF beta 2 Antibody, Novus Biologicals (NBP1-36573)
Rabbit Polyclonal TGF beta 2 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: TGF beta 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR