Rabbit Polyclonal anti-TGF alpha Antibody, Novus Biologicals (NBP1-02694)

NBP1-02694
102155-514EA 466.15 USD
102155-514
Rabbit Polyclonal anti-TGF alpha Antibody, Novus Biologicals (NBP1-02694)
Rabbit Polyclonal TGF alpha Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: TGF alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR