Anti-TFIIE-alpha Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46196
102175-780EA 635.51 USD
102175-780
Anti-TFIIE-alpha Rabbit Polyclonal Antibody
Antibodies
The TFIIE-alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to TFIIE-alpha. This antibody reacts with human, mouse. The TFIIE-alpha Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation.

Type: Primary
Antigen: TFIIE-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR