Anti-TFEB Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-30908
102163-860EA 349 USD
102163-860
Anti-TFEB Rabbit Polyclonal Antibody
The TFEB Antibody from Novus Biologicals is a rabbit polyclonal antibody to TFEB. This antibody reacts with human. The TFEB Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR