Anti-TFAP4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46202
102175-804EA 635.51 USD
102175-804
Anti-TFAP4 Rabbit Polyclonal Antibody
Antibodies
The TFAP4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TFAP4. This antibody reacts with human. The TFAP4 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TFAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR