Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TET1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78965 NBP1-78965SS
The TET1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TET1. This antibody reacts with mouse. The TET1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC / IF (Negative).

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: TET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.025 ml NBP1-78965SS 102213-562 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

100 μl NBP1-78965 102213-560 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TET1
Antigen Name Tet Methylcytosine Dioxygenase 1
Reactivity Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms ten-eleven translocation-1, CXXC6FLJ10839, Ten-eleven translocation 1 gene protein, methylcytosine dioxygenase TET1, LCXEC 1.14.11.n2, FLJ41442, tet oncogene 1, KIAA1676CXXC-type zinc finger protein 6, Leukemia-associated protein with a CXXC domain, bA119F7.1, CXXC6, CXXC finger 6, LCX CXXC6FLJ10839, methylcytosine dioxygenase TET1, LCXEC 1.14.11.n2, CXXC6, CXXC finger 6, tet oncogene 1, ten-eleven translocation-1, KIAA1676CXXC-type zinc finger protein 6, LCX, bA119F7.1, FLJ41442, Ten-eleven translocation 1 gene protein, Leukemia-associated protein with a CXXC domain
Gene ID 80312
Concentration 1.07 mg/ml
Immunogen A synthetic peptide made to an internal portion of the human TET1 protein (between residues 170-800) [UniProt Q8NFU7]
Purification Immunogen affinity purified
Storage Buffer PBS and 30% Glycerol
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty