Rabbit Polyclonal anti-TENC1 Antibody, Novus Biologicals (NBP2-13028)

NBP2-13028
102252-836EA 466.15 USD
102252-836
Rabbit Polyclonal anti-TENC1 Antibody, Novus Biologicals (NBP2-13028)
Rabbit Polyclonal TENC1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: TENC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR