Anti-Tenascin C Rabbit Polyclonal Antibody

41720002
102103-946EA 423.08 USD
102103-946
Anti-Tenascin C Rabbit Polyclonal Antibody
Rabbit Polyclonal Tenascin C Antibody. Tested Applications: ELISA. Tested Reactivity: Human.

Type: Primary Antibodies
Antigen: Tenascin C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR