Rabbit Polyclonal anti-Tenascin C Antibody, Novus Biologicals (41660002)

41660002
102103-938EA 423.08 USD
102103-938
Rabbit Polyclonal anti-Tenascin C Antibody, Novus Biologicals (41660002)
Rabbit Polyclonal Tenascin C Antibody. Tested Applications: ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: Tenascin C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR