Anti-TEM8/ANTXR1 Rabbit Polyclonal Antibody

NBP1-45884
102175-302EA 466.15 USD
102175-302
Anti-TEM8/ANTXR1 Rabbit Polyclonal Antibody
The TEM8 / ANTXR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TEM8 / ANTXR1. This antibody reacts with human, mouse, rat. The TEM8 / ANTXR1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TEM8/ANTXR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR