Anti-TEM7/PLXDC1 Rabbit Polyclonal Antibody

NBP1-76832
102209-540EA 466.15 USD
102209-540
Anti-TEM7/PLXDC1 Rabbit Polyclonal Antibody
The TEM7 / PLXDC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TEM7 / PLXDC1. This antibody reacts with human, mouse, rat. The TEM7 / PLXDC1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 2.5 ?g/ml

Type: Primary
Antigen: TEM7/PLXDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR