Anti-CCL25/TECK Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-40993
102169-714EA 564.74 USD
102169-714
Anti-CCL25/TECK Rabbit Polyclonal Antibody
Antibodies
The CCL25 / TECK Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCL25 / TECK. This antibody reacts with human, mouse. The CCL25 / TECK Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Functional.

Type: Primary
Antigen: CCL25/TECK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR