Anti-TEAD4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-82386
102216-832EA 564.74 USD
102216-832
Anti-TEAD4 Rabbit Polyclonal Antibody
Antibodies
The TEAD4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TEAD4. This antibody reacts with human, mouse. The TEAD4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TEAD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR