Anti-TDRD7 Rabbit Polyclonal Antibody

30640002
102103-274EA 466.15 USD
102103-274
Anti-TDRD7 Rabbit Polyclonal Antibody
Rabbit Polyclonal TDRD7 Antibody. Tested Applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: TDRD7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR