Anti-TDO2 Rabbit Polyclonal Antibody

NBP1-54877
102184-714EA 493.85 USD
102184-714
Anti-TDO2 Rabbit Polyclonal Antibody
The TDO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TDO2. This antibody reacts with human. The TDO2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TDO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR