Anti-TCL1A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54693
102184-352EA 564.74 USD
102184-352
Anti-TCL1A Rabbit Polyclonal Antibody
Antibodies
The TCL1A Antibody from Novus Biologicals is a rabbit polyclonal antibody to TCL1A. This antibody reacts with human. The TCL1A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TCL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR