Anti-TCIRG1 Rabbit Polyclonal Antibody

43480002
102104-014EA 521.54 USD
102104-014
Anti-TCIRG1 Rabbit Polyclonal Antibody
Rabbit Polyclonal TCIRG1 Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary Antibodies
Antigen: TCIRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR