Anti-TCF-3/E2A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77119
102210-664EA 564.74 USD
102210-664
Anti-TCF-3/E2A Rabbit Polyclonal Antibody
Antibodies
The TCF-3 / E2A Antibody from Novus Biologicals is a rabbit polyclonal antibody to TCF-3 / E2A. This antibody reacts with human, mouse, rat. The TCF-3 / E2A Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: TCF-3/E2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR