Anti-TCEA1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-26625
102161-102EA 635.51 USD
102161-102
Anti-TCEA1 Rabbit Polyclonal Antibody
Antibodies
The TCEA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TCEA1. This antibody reacts with human, mouse. The TCEA1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation (Negative).

Type: Primary
Antigen: TCEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR