Anti-TBX4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32879
102166-494EA 635.51 USD
102166-494
Anti-TBX4 Rabbit Polyclonal Antibody
Antibodies
The TBX4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBX4. This antibody reacts with human. The TBX4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR