Anti-TBCD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79913
102215-474EA 564.74 USD
102215-474
Anti-TBCD Rabbit Polyclonal Antibody
Antibodies
The TBCD Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBCD. This antibody reacts with human. The TBCD Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TBCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR