Anti-TBC1D2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79028
102213-734EA 635.51 USD
102213-734
Anti-TBC1D2 Rabbit Polyclonal Antibody
Antibodies
The TBC1D2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBC1D2. This antibody reacts with human. The TBC1D2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TBC1D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR