Anti-TBC1D10A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79843
102215-334EA 564.74 USD
102215-334
Anti-TBC1D10A Rabbit Polyclonal Antibody
Antibodies
The TBC1D10A Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBC1D10A. This antibody reacts with human. The TBC1D10A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TBC1D10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR