Anti-TAZ/WWTR1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB600-220
102133-514EA 491.08 USD
102133-514
Anti-TAZ/WWTR1 Rabbit Polyclonal Antibody
Antibodies
The TAZ / WWTR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TAZ / WWTR1. This antibody reacts with human, mouse. The TAZ / WWTR1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TAZ/WWTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR